High Visibility Caps, Elysee®, Roman / Daniel
Supplier: FELDTMANN HELMUT
Certificates
About this item
Made from 80% polyester / 20% cotton.
- 6-part top with embroidered air holes
- Integrated reflective piping
- 2-colour peak with embroidered intermediate part (sandwich-peak)
- Universal size, adjustable width
Specifications
- Catalog No:
- 113-08325
- 113-08326
- Risk area:
- HiVis
- HiVis
- Gender:
- Unisex
- Unisex
- Fabric:
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton