VWR®, Yeast extract
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
- High purity extract
- Used for supplementing media
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.