Herbicide reference standard, Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea) 1000 µg/ml in acetonitrile
Catalog # PROOU-EPA-1188
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:1746-81-2
- Description:Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea) 1,000 µg/ml in acetonitrile
- Formula:C₉H₁₁ClN₂O₂
- Description:Monolinuro