Herbicide reference standard, Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 1000 µg/ml in hexane
PROOU-EPA-1182
:
- Pk:1
- CAS No.:330-55-2
- Description:Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 1,000 µg/ml in hexane
- Formula:C₉H₁₀Cl₂N₂O₂
- Description:Linuro