ContactUsLinkComponent
Tin standard solution, 10000 mg/L Sn in water standard for AAS
  PROOU-IAA-050
 :  
Tin standard solution, 10000 mg/L Sn in water standard for AAS
  PROOU-IAA-050
 :  
  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Element:
    Tin
  • Symbol:
    Sn