ContactUsLinkComponent
Phosphorus standard solution, 1,000 mg/L P in dilute nitric acid standard for AAS
Phosphorus standard solution, 1,000 mg/L P in dilute nitric acid standard for AAS
  PROOU-IAA-215
 :  
Phosphorus standard solution, 1,000 mg/L P in dilute nitric acid standard for AAS
  PROOU-IAA-215
 :  
  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Phosphorus
  • Symbol:
    P