Organic reference standard, p-Terphenyl 2,000 µg/ml in dichloromethane
Catalog # PROOU-IST-490-1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:92-94-4
- Description:p-Terphenyl 2,000 µg/ml in dichloromethane
- Formula:C₁₈H₁₄
- Description:p-Terphenyl