Samarium standard solution, 10,000 mg/L Sm in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-062
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:10000mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:10 mL
- Element:Samarium
- Symbol:Sm