Ruthenium standard solution, 1,000 mg/L Ru in 5% hydrochloric acid standard for AAS
Catalog # PROOU-IAA-244
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:5% HCl
- Pack type:Glass bottle
- Pk:125 mL
- Element:Ruthenium
- Symbol:Ru