Rhodium standard solution, 1000 mg/L Rh in 5% hydrochloric acid standard for ICP
Catalog # PROOU-ICP-045
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:5% HCl
- Pack type:Glass bottle
- Pk:125 mL
- Element:Rhodium
- Symbol:Rh