Pesticide reference standard, δ-BHC 1,000 µg/ml in methanol
Catalog # PROOU-EPA-1078
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:319-86-8
- Description:δ-BHC 1,000 µg/ml in methanol
- Formula:C₆H₆Cl₆
- Description:δ-BHC