ContactUsLinkComponent
Terbium standard solution, 10000 mg/L Tb in dil. nitric acid standard for ICP
  PROOU-ICP-165
 :  
Terbium standard solution, 10000 mg/L Tb in dil. nitric acid standard for ICP
  PROOU-ICP-165
 :  
  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Terbium
  • Symbol:
    Tb