Order Entry
Slovenia
ContactUsLinkComponent
 
Selenium standard solution, 1,000 mg/L Se in dil. nitric acid standard for AAS
  PROOU-IAA-234
 :  
Selenium standard solution, 1,000 mg/L Se in dil. nitric acid standard for AAS
  PROOU-IAA-234
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Selenium
  • Symbol:
    Se