Order Entry
Slovenia
ContactUsLinkComponent
 
Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid standard for ICP
  PROOU-ICP-021
 :  
Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid standard for ICP
  PROOU-ICP-021
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Scandium
  • Symbol:
    Sc