Order Entry
Slovenia
ContactUsLinkComponent
Iron standard solution, 10000 mg/L Fe in dil. nitric acid standard for AAS
Iron standard solution, 10000 mg/L Fe in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-026
CAS Number:  
Iron standard solution, 10000 mg/L Fe in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-026
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item