ContactUsLinkComponent
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for AAS
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for AAS
  PROOU-IAA-039
 :  
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for AAS
  PROOU-IAA-039
 :  
  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Element:
    Yttrium
  • Symbol:
    Y