ContactUsLinkComponent
Pipette aid, SP Bel-Art
Pipette aid, SP Bel-Art
Catalog # BELAF378860000
Pipette aid, SP Bel-Art
Catalog # BELAF378860000
Specifications
  • Colour:
    Blue/white
  • Description:
    Pipette aid for pipettes, Vikem® viny bulb with PE chuck, Hר: 100×50 mm
  • Seq:
    02
  • Pk:
    6
Specifications
About this item
This pipette aid is used by squeezing, then releasing the bulb to aspirate fluid into the pipette.
  • Quickly cover pipette top with a gloved finger to control and dispense fluid
  • Vikem® vinyl bulb has a tapered polyethylene chuck for quick removal when pipette is filled