ContactUsLinkComponent
Electrofusion buffer, BTXpress Cytofusion® Medium C
Electrofusion buffer, BTXpress Cytofusion® Medium C
Catalog # 731-0348
Supplier:  BTX
CAS Number:  
Electrofusion buffer, BTXpress Cytofusion® Medium C
Catalog # 731-0348
Supplier:  BTX
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Application:
  • Description:
    Electrofusion buffer, BTXpress Cytofusion® Medium C
  • Environmentally Preferable:
Specifications
About this item
BTXpress Cytofusion® Medium C is an advanced electrofusion buffer designed especially for hybridoma production. The low conductivity buffer is specially formulated to minimise cell turbulence during cell alignment and heating during electrofusion, for robust cell fusion efficiency and high cell viability. BTXpress Cytofusion® Medium C is sterile filtered from the highest quality non animal, medical grade reagents.