Phosphorus standard solution, 1,000 mg/L P in water (from (NH₄)PO₄H₂) standard for ICP
Frequently Bought Together
Certified colour reference standards
: Lovibond Tintometer
Riboflavine (vitamin B2)
: EDQM
2-Ethylcaproic acid
: EHRENSTORFER
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
: EHRENSTORFER
Gold standard solution, for ICP, in 20% HCl, VHG
: LGC Standards PROMOCHEM
Buffer Solutions
: Thermo Fisher Scientific
VWR®, Flow Cup viscosity standards
: VWR Chemicals
Fonofos certified 1 * 0,1 g
: EHRENSTORFER
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
: YSI ENVIRONMENTAL