Cadmium standard solution, 10000 mg/L Cd in dil. nitric acid standard for ICP
Catalog # PROOU-ICP-148
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:125 mL
- Element:Cadmium
- Symbol:Cd