Orders LinkContactUsLinkComponent
Oil sample bottle, with cork, WHEATON®
Oil sample bottle, with cork, WHEATON®
125 ml, 37 mm, 163 mm
Catalog # WHEAW216994
Supplier:   DWK Life Sciences
Oil sample bottle, with cork, WHEATON®
Catalog # WHEAW216994
Supplier:  DWK Life Sciences
Oil sample bottle, with cork, WHEATON®
Oil sample bottle, with cork, WHEATON®
125 ml, 37 mm, 163 mm
Price
485,00 €
Log in for availability
1 * 144 items
Product Details & Documents
Clear, Type III soda-lime glass.
Bottles come with cork inserted.
Specifications
  • Ext.Ø:
    37 mm
  • Capacity:
    125 ml
  • Height:
    163 mm