ABsolute™ qPCR Low ROX Mixes
ABsolute™ qPCR low ROX mixes are optimised for use with real time instruments that are able to detect long wavelength dyes with greater sensitivity, such as the ABI 7500. The lower level of ROX still facilitates normalisation across the plate, while maintaining the sensitivity of the assay. The mixes contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X qPCR mix contains a proprietary reaction buffer that provides high sensitivity, specificity and reproducibility.
- Contains hot start enzyme Thermo-Start® DNA polymerase for increased specificity and optimised buffer for maximum sensitivity
- Excellent reproducibility, with all batches undergoing extensive quality control to ensure batch-to-batch and assay-to-assay consistency
- Optimised buffer specifically designed for real time instruments that are able to detect long wavelength dyes with greater sensitivity
- Application:qPCR dye
Frequently Bought Together