VWR® Dip Slides
Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.
- Supplied sterile
- Sampling takes only a few seconds
- Results are easily read after just 24 to 48 hours
- Paddle size:80×28
- Contact area:10
- Dimensions:30 mm tube, 110 mm high
- Tray W×L:50x22
- Type:Push cap
25811725.pdf SDS_SL_SI_VWRC535092Q.pdf SDS_SL_SI_VWRC53510.pdf SDS_SL_SI_VWRC53511.pdf SDS_SL_SI_VWRC53512.pdf
Frequently Bought Together