Accessories for dissolved oxygen probes
: WTW
Frequently Bought Together
JUG 1000ML 31,5MM GREEN GLAS 1 * 1.183 items
: PONT PACKAGING
Accessories for PURELAB Flex Systems
: ELGA LabWater
pH indicator test strips, PEHANON®
: MACHEREY-NAGEL
Stirring rods
: DWK Life Sciences
VWR®, Overalls with Hood or Collar
: VWR Collection
Membrane filters, track etched
: Sartorius
Sulphuric acid 25%, GPR RECTAPUR®
: VWR Chemicals
Ammonium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
BLANK NUT LONG 1 * 1 items
: Agilent