Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Reagents for ion specific colorimeters
Reagents for ion specific colorimeters
Reagents for ion specific colorimeters
Catalog # 662-0019
Supplier:
Hanna
CAS Number:
Expand image
Reagents for ion specific colorimeters
Catalog # 662-0019
Supplier:
Hanna
CAS Number:
Specifications
Pk:
100 Tests
Description:
Ammonia high range
Range:
0,0 - 50,0 mg/l
Specifications
About this item
Product Details
Documentation
Product Details
Documentation