Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lights
UV Lights, Lamps and Bulbs
UV multiple ray lamp
UV multiple ray lamp
UV multiple ray lamp
UVPA95-0313-02
:
UVP ULTRA VIOLET PRODUCTS
UV multiple ray lamp
UVPA95-0313-02
:
UVP ULTRA VIOLET PRODUCTS
For:
Benchtop use
Description:
Multiple ray UV lamp, three height positions, 365/254 nm, cool white/north sky light, 230 V
Environmentally Preferable:
Model:
MLR-58
Output:
8 W
Pk:
1
Wavelength:
365/254 nm