Precision probes for 735
: Testo
Product Family Options
- DescriptionRangeAccuracyØ×L
- TC type T flexible air probe with PTFE cable−50…+250 °C±0,3 °C (−20...+70 °C)
±(0,7 °C (±0,5% mv)1,5×2000 mm1 * 1 items50,00 € - PT100 glass-coated laboratory probe with replaceable glass pipe−50…+400 °C±0,2 °C
±0,2% mv (+200...+800 °C)6×200 mm1 * 1 items134,00 € - Highly accurate immersion PT100 probe with certificate−40…+300 °C±0,3 °C (−80... −40 °C) ;
±0,1 °C + 0,05% mv (−40...0 °C)
±0,05 °C (0 ...100 °C)
±0,05 °C + 0,05 % mv (+100...+300 °C)4×295 mm1 * 1 items472,00 €