Orders LinkContactUsLinkComponent
Thermometer probes
Supplier:   COMARK
Thermometer probes
Supplier:  COMARK
Certificates
Thermometer probes
8 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Range
    Application
    Ø×L
    Availability
    Price
  • 620-0333
    Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M
    Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M
    −100…+850 °C
    General purpose air
    1,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 310,00 €
    310,00 €
    1 * 1 items
       310,00 €
  • 620-3174
    Flexible wire type K air probe, with sub-miniature connector, AK28M
    Flexible wire type K air probe, with sub-miniature connector, AK28M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 76,90 €
    76,90 €
    1 * 1 items
       76,90 €
  • COMAAK29M
    Flexible wire type K air probe, with sub-miniature connector, AK29M
    Flexible wire type K air probe, with sub-miniature connector, AK29M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 80,50 €
    80,50 €
    1 * 1 items
       80,50 €
  • COMAAK31M
    Flexible wire type K air probe, with sub-miniature connector, AK31M
    Flexible wire type K air probe, with sub-miniature connector, AK31M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 68,50 €
    68,50 €
    1 * 1 items
       68,50 €
  • 620-0334
    Type T high temperature air probe up to +400 °C, sub-miniature connector, AK33M
    Type T high temperature air probe up to +400 °C, sub-miniature connector, AK33M
    −100…+400 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 66,90 €
    66,90 €
    1 * 1 items
       66,90 €
  • 620-3171
    Type K industrial probe, with sub-miniature connector, IK21M
    Type K industrial probe, with sub-miniature connector, IK21M
    −100...+850 °C
    Immersion
    1,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 272,00 €
    272,00 €
    1 * 1 items
       272,00 €
  • 620-3172
    Fast response type K probe, sub-miniature connector, PK24M
    Fast response type K probe, sub-miniature connector, PK24M
    −50…+250 °C
    Penetration
    3,3×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 124,00 €
    124,00 €
    1 * 1 items
       124,00 €
  • 620-3173
    Fast response type K probe for up to +250 °C, subminiature connector, SK21M
    Fast response type K probe for up to +250 °C, subminiature connector, SK21M
    −50...+250 °C
    Surface
    2,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 427,00 €
    427,00 €
    1 * 1 items
       427,00 €