Orders LinkContactUsLinkComponent
Accessories for empty solvent resistant columns
Accessories for empty solvent resistant columns
Capillary tubing, PTFE, 1,1 mm ID, 1,8 mm OD, 5 m length
Catalog # 19-0041-01
Supplier:   Cytiva
Accessories for empty solvent resistant columns
Catalog # 19-0041-01
Supplier:  Cytiva
Accessories for empty solvent resistant columns
Accessories for empty solvent resistant columns
Capillary tubing, PTFE, 1,1 mm ID, 1,8 mm OD, 5 m length
Price
161,00 €
Log in for availability
1 * 5 m
Product Details & Documents
Specifications
  • Description:
    Capillary tubing, PTFE, 1,1 mm ID, 1,8 mm OD, 5 m length