ContactUsLinkComponent
Protein G Sepharose™ 4 Fast Flow Affinity Chromatography Media
 :  Cytiva
undefined
Protein G Sepharose™ 4 Fast Flow Affinity Chromatography Media
 :  Cytiva
  • pH range:
    3 - 9
  • Matrix:
    Cross-linked agarose, 4%, spherical
  • Ligand:
    Recombinant protein G lacking albumin-binding region, produced in E. coli
  • Storage temperature:
    2 - 8 °C
  • Ligand density:
    2 mg Protein G/ml
  • Ligand coupling method:
    Cyanogen bromide activation
Frequently Bought Together