Orders LinkContactUsLinkComponent
Detectable sampling scoops, Sterileware®, Sense-able Scoops®, SP Bel-Art
Detectable sampling scoops, Sterileware®, Sense-able Scoops®, SP Bel-Art
Detectable sampling scoops, Sterileware®, Sense-able Scoops®, SP Bel-Art
Certificates
Detectable sampling scoops, Sterileware®, Sense-able Scoops®, SP Bel-Art
Detectable sampling scoops, Sterileware®, Sense-able Scoops®, SP Bel-Art
3 Options Available Below
Product Details & Documents
PS, blue. These scoops are made of plastic impregnated with stainless steel powder which permits fragments as small as 1,6 mm³ to be detected with standard metal detection and X-ray equipment.
  • Stainless steel powder homogeneously incorporated into scoops from top to bottom
  • FDA grade materials safe for food, drug and cosmetic contact
  • Long handle increases the distance between the hand and sample to reduce contamination and exposure potential (60 ml and 125 ml models)
  • Flat bottom allows scoops to remain upright for use as weighing boats or to rest on a flat surface without spilling
  • Sterilised by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilisation
The blue colouring conforms to industry recognised standards for metal detectable materials and provides a contrasting colour to most sampled products.
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Width
    Material
    Colour
    Availability
    Price
  • 60 ml
    203 mm
    38 mm
    PS
    Blue
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 532,00 €
    532,00 €
    1 * 100 items
       532,00 €
  • 125 ml
    218 mm
    46 mm
    PS
    Blue
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 795,00 €
    795,00 €
    1 * 100 items
       795,00 €
  • 250 ml
    191 mm
    51 mm
    PS
    Blue
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 1 380,00 €
    1 380,00 €
    1 * 100 items
       1 380,00 €