dNTP mix
Catalog # 733-1271
Supplier: Quantabio
CAS Number:
Specifications
- Pk:1.000 µl
- Application:
- Description:dNTP Mix
- Seq:01
- Environmentally Preferable:
Specifications
About this item
The 10 mM dNTP mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (qPCR). It is suitable for use in conventional end-point PCR, real time qPCR, first-strand cDNA synthesis, as well as other applications that require dNTP as substrate. The use of qPCR certified dNTPs is a simple but important measure to ensure consistent assay performance.