Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
Supplier:   LAUDA
Lifting platforms for Lauda baths
Supplier:  LAUDA
Certificates
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
3 Options Available Below
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 461-2948
    Lifting platform, for 1× E 4/E 103/E 203; 2× A 12 and 6× A 24
    Lifting platform, for 1× E 4/E 103/E 203; 2× A 12 and 6× A 24
    115×70 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 406,00 €
    406,00 €
    1 * 1 items
       406,00 €
  • 461-2951
    Lifting platform, for 1× E 106 T/E 206 T/ET 6 and 2× A 24
    Lifting platform, for 1× E 106 T/E 206 T/ET 6 and 2× A 24
    110×260 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 414,00 €
    414,00 €
    1 * 1 items
       414,00 €
  • 461-2909
    Lifting platform, for 1× E 10/E 15/ET 12/E 112 T/E 212 T/E 312/E 111/E 211/ET 12/ET 20 and 2× A 24/E 20/E 25/E 120 T/E 220 T/ET 20/E 119/E 219/E 125/E 225/E 320/E 326
    Lifting platform, for 1× E 10/E 15/ET 12/E 112 T/E 212 T/E 312/E 111/E 211/ET 12/ET 20 and 2× A 24/E 20/E 25/E 120 T/E 220 T/ET 20/E 119/E 219/E 125/E 225/E 320/E 326
    140×270 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 414,00 €
    414,00 €
    1 * 1 items
       414,00 €