Interchangeable blocks for HLC thermomixers and block heaters
Supplier: HAEP LABOR
Certificates
About this item
High capacity blocks made from anodised aluminium, all blocks have W×D of 128×85 mm. They are simply plugged into the block heaters and screwed together.
Delivery: Supplied with block extraction tool.
Frequently Bought Together