ContactUsLinkComponent
NeuroSlide™ 21 storage system
NeuroSlide™ 21 storage system
  720-2351
 :  CELLPATH
undefined
NeuroSlide™ 21 storage system
  720-2351
 :  CELLPATH
  • No. of slides:
    21
  • W×D:
    260×380  mm
  • Colour:
    Transparent
  • Description:
    Slide holders
  • Material:
    Plastic
  • Pk:
    10
21 compartments for 76×26, 50×76 and 76×76 mm slides or 35 mm transparencies with hanging system for storage in conventional filing cabinet.