Grease, stopcocks, silicone-free
Grease for lubricating burette stopcocks, silicone-free, high vacuum quality, 30 g
Catalog # BRND61610
Supplier: Brand
Jump to:
Product Details & Documents
Ideal for lubricating burette stopcocks.
- Easy to remove
- Good acid- and base-resistance
- Operating temperature −40 to +320 °C
Specifications
- Description:Grease for lubricating burette stopcocks, silicone-free, high vacuum quality, 30 g