Order Entry
Slovenia
ContactUsLinkComponent
 
Pesticide reference standard, 1,2,3-Trichlorobenzene 100 µg/ml in methanol
  PROOU-HC-420-1
 :  
Pesticide reference standard, 1,2,3-Trichlorobenzene 100 µg/ml in methanol
  PROOU-HC-420-1
 :  

 

  • Pk:
    1
  • CAS No.:
    87-61-6
  • Description:
    1,2,3-Trichlorobenzene 100 µg/ml in methanol
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,2,3-Trichlorobenzene