2-Chlorophenol
Glass bottle for solids, 100 mg
Catalog # ACCUA-013
Supplier: AccuStandard
Jump to:
Product Details & Documents
Synonyms: 1-Chloro-2-hydroxybenzene
- CAS Number:95-57-8
- Formula:C₆H₅ClO
- MDL Number:MFCD00002159
- Molecular Weight:128.56 g/mol
- Merck Index:12,02173
- Boiling Pt:175 °C (1013 hPa)
- Melting Pt:7…8 °C
- Density:1.25 g/cm³ (20 °C)
- Flash Pt:72 °C
- Storage Temperature:Ambient
- EINECS:202-433-2
- UN:2021
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



