Orders LinkContactUsLinkComponent
Cryoboxes
Cryoboxes
Supplier:   RATIOLAB
Cryoboxes
Supplier:  RATIOLAB
Certificates
Cryoboxes
2 Options Available Below
Product Details & Documents
Cryoboxes, cardboard, for storage of cryovials at low temperatures.
  • Waterproof coating
  • Dividers must to be ordered separately
  • Available in different heights and five colours
Product Family Options
Cryoboxes
Product Information
  • Compartments
    For
    Colour
    W×D×H ext.
    Availability
    Price
  • -
    -
    White
    75×75×50 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 34,90 €
    34,90 €
    1 * 10 items
       34,90 €
Dividers
Product Information
  • Compartments
    For
    Colour
    W×D×H ext.
    Availability
    Price
  • 5×5
    Tubes 11,0 mm Ø
    White
    68×68×30 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 16,20 €
    16,20 €
    1 * 10 items
       16,20 €