Syringes for HPLC autosamplers, Thermo Scientific instruments
Catalog # 548-1644
Supplier: Trajan Scientific and Medical
Specifications
- For:AS100/300 and AS1000/3000/3500 LC autosamplers
- Type:250C-THERMOSPEC
- Volume:250 µl
- Description:Gas-tight, with PTFE-tipped plunger, 1/4-28 UNF front thread
- Environmentally Preferable:
- Pk:1