Order Entry
Slovenia
ContactUsLinkComponent
Organic reference standard, (±)-2,3-Dibromopropionic acid 1000 µg/ml in tert-butyl methyl ether
Organic reference standard, (±)-2,3-Dibromopropionic acid 1000 µg/ml in tert-butyl methyl ether
Catalog # PROOU-PPS-390-1
CAS Number:  
Organic reference standard, (±)-2,3-Dibromopropionic acid 1000 µg/ml in tert-butyl methyl ether
Catalog # PROOU-PPS-390-1
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    600-05-5
  • Description:
    (±)-2,3-Dibromopropionic acid 1,000 µg/ml in tert-butyl methyl ether
  • Formula:
    C₃H₄Br₂O₂
  • Description:
    (±)-2,3-Dibromopropionic acid

Specifications

About this item