2-Adamantanone
Catalog # MOLE23794765-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:150.22 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:43 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-847-2
CAS Number:700-58-3
Formula:C₁₀H₁₄O
MDL Number:MFCD00074737