ContactUsLinkComponent
Organic reference standard, Propylamine 1,000 µg/ml in methanol
Organic reference standard, Propylamine 1,000 µg/ml in methanol
Catalog # PROOU-EPA-1232
CAS Number:  
Organic reference standard, Propylamine 1,000 µg/ml in methanol
Catalog # PROOU-EPA-1232
CAS Number:  
Specifications
  • Pk:
    1
  • CAS No.:
    107-10-8
  • Description:
    Propylamine 1,000 µg/ml in methanol
  • Formula:
    C₃H₉N
  • Description:
    Propylamine