Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Herbicide reference standard, 2-Picoline 5000 µg/ml in methanol
Herbicide reference standard, 2-Picoline 5000 µg/ml in methanol
PROOU-EPA-1156
:
LGC Standards PROMOCHEM
:
Herbicide reference standard, 2-Picoline 5000 µg/ml in methanol
PROOU-EPA-1156
:
LGC Standards PROMOCHEM
:
Pk:
1
CAS No.:
109-06-8
Description:
2-Picoline 5,000 µg/ml in methanol
Formula:
C₆H₇N
Description:
2-Picolin