Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Supplier:   MARIENFELD
Capillary tubes
Supplier:  MARIENFELD
Certificates
Capillary tubes
2 Options Available Below
Product Details & Documents
Glass, clear.
  • Na-heparinized (coated)
Capillaries, Na-heparinized (coated)
For blood gas analysis.
Product Family Options
Product Information
  • Length
    Ø
    Volume
    Availability
    Price
  • MARI2951300
    Capillary Tubes
    Capillary Tubes
    75 mm
    2,3 mm
    130 µl
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • MARI2951303
    Capillary Tubes
    Capillary Tubes
    75 mm
    1,75 mm
    82,5 µl
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Closure cap for blood gas capillary tubes with Ø 1,75 mm
    Closure cap for blood gas capillary tubes with Ø 1,75 mm
    MARI2960414
    Log in for availability
    1 * 500 items