Orders LinkContactUsLinkComponent
Slide folder, 'Munich'
Slide folder, 'Munich'
Preparation pasteboards with cover, Natural, Cardboard, 24
Catalog # MARI6001313
Supplier:   MARIENFELD
Slide folder, 'Munich'
Catalog # MARI6001313
Supplier:  MARIENFELD
Slide folder, 'Munich'
Slide folder, 'Munich'
Preparation pasteboards with cover, Natural, Cardboard, 24
Price
78,56 €
Log in for availability
1 * 10 items
Product Details & Documents
Cardboard base with three compartments (78×220 mm).
  • Each compartment can accomodate up to eight slides
  • Hinged cover with three windows (56×200 mm)
Specifications
  • No. of slides:
    24
  • Colour:
    Natural
  • Description:
    Preparation pasteboards with cover
  • Material:
    Cardboard