Disodium hydrogen phosphate
Supplier: Roth Carl
Jump to:
About this item
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
- CAS Number:7558-79-4
- Formula:Na₂HPO₄
- MDL Number:MFCD00003496
- Molecular Weight:141.96 g/mol
- Merck Index:12,08805
- Melting Pt:250 °C
- Density:1.53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-448-7
- REACH:01-2119489797-11
2 Options Available Below
Product Details & Documents
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
- CAS Number:7558-79-4
- Formula:Na₂HPO₄
- MDL Number:MFCD00003496
- Molecular Weight:141.96 g/mol
- Merck Index:12,08805
- Melting Pt:250 °C
- Density:1.53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-448-7
- REACH:01-2119489797-11
Product Family Options
Product Information
- Pack typePkAvailabilityPrice