Orders LinkContactUsLinkComponent
Microscopy slides
Microscopy slides
Microscopy slides with clipped corners and 12 blue rings (Ø 13/14 mm)
Catalog # MARI1613100
Supplier:   MARIENFELD
Microscopy slides
Catalog # MARI1613100
Supplier:  MARIENFELD
Microscopy slides
Microscopy slides
Microscopy slides with clipped corners and 12 blue rings (Ø 13/14 mm)
Price
71,87 €
Log in for availability
1 * 10 items
Product Details & Documents
These microscope slides feature highly visible blue rings (Ø 14mm).
Documents
Specifications
  • Description:
    Microscopy slides with clipped corners and 12 blue rings (Ø 13/14 mm)