Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Labels
Slide labels with accession number
Slide labels with accession number
Slide labels with accession number
Catalog # TIMECVR-58
Supplier:
PRECISION DYNAMICS TIMEM
Expand image
Slide labels with accession number
Catalog # TIMECVR-58
Supplier:
PRECISION DYNAMICS TIMEM
Specifications
Colour:
Black on white
Description:
Overlaminated slide labels, accession numbered
Dimensions:
22×16 mm
Packed:
On roll
Pk:
1.000
Specifications
About this item
Product Details
Product Details