Orders LinkContactUsLinkComponent
Sodium nitrate
Sodium nitrate
Oxidizing
Irritant
Supplier:   Roth Carl
Sodium nitrate
Supplier:  Roth Carl
Certificates
3 Options Available Below
Product Details & Documents
  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:13,08720
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:308 °C
  • Density:2.26 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41
Product Family Options
Product Information
  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 28,60 €
    28,60 €
    1 * 1 kg
       28,60 €
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg
  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg